Anti-MIP3B Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
M1202-65L-PE
USBIM1202-65L-PEEA 677 EUR
USBIM1202-65L-PE
Anti-MIP3B Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: MIP3B
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR