Anti-Listeria sp. Rabbit Polyclonal Antibody (FITC (Fluorescein))

Toimittaja: US Biological
L2650-09C
USBIL2650-09CEA 435 EUR
USBIL2650-09C
Anti-Listeria sp. Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Recommended dilution: Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: Listeria sp.
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR