Anti-L11 Ribosomal Protein Mouse Monoclonal Antibody [clone: 6A104]

Toimittaja: US Biological
L0011
USBIL0011EA 809 EUR
USBIL0011
Anti-L11 Ribosomal Protein Mouse Monoclonal Antibody [clone: 6A104]
Vasta-aineet
Recommended dilution: Western Blot: 1-3?g/ml

Type: Primary
Antigen: L11 Ribosomal Protein
Clonality: monoclonal
Clone: 6A104
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR