Anti-Interferon Stimulating Gene 15 Mouse Monoclonal Antibody [clone: 7G32]

Toimittaja: US Biological
I7662-94B-100
USBII7662-94B-100EA 608 EUR
USBII7662-94B-100
Anti-Interferon Stimulating Gene 15 Mouse Monoclonal Antibody [clone: 7G32]
Vasta-aineet
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Interferon Stimulating Gene 15
Clonality: Monoclonal
Clone: 7G32
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR