Anti-HLA Class 2 Antigen DP, DQ, DR Mouse Monoclonal Antibody [clone: 10D177]

Toimittaja: US Biological
H6099-85E
USBIH6099-85EEA 660 EUR
USBIH6099-85E
Anti-HLA Class 2 Antigen DP, DQ, DR Mouse Monoclonal Antibody [clone: 10D177]
Vasta-aineet
Recommended dilution: Flow Cytometry: 1:100-1:200;

Type: Primary
Antigen: HLA Class 2 Antigen DP, DQ, DR
Clonality: monoclonal
Clone: 10D177
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR