Anti-Hexokinase, Type IV Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Toimittaja: US Biological
H2040-51E-AP
USBIH2040-51E-APEA 884 EUR
USBIH2040-51E-AP
Anti-Hexokinase, Type IV Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Hexokinase, Type IV
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR