Anti-HK1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
H2035-25C-PE
USBIH2035-25C-PEEA 884 EUR
USBIH2035-25C-PE
Anti-HK1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: HK1 (hexokinase 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR