Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Rabbit polyclonal antibody

Toimittaja: US Biological
G8140-16K
USBIG8140-16KEA 682 EUR
USBIG8140-16K
Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Rabbit polyclonal antibody
Vasta-aineet
Type: Primary
Antigen: Glyceraldehyde 3-Phosphate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR