Anti-CFB Mouse monoclonal antibody [clone: 10K167]

Toimittaja: US Biological
C7850-60B
USBIC7850-60BEA 539 EUR
USBIC7850-60B
Anti-CFB Mouse monoclonal antibody [clone: 10K167]
Vasta-aineet
Type: Primary
Antigen: CFB (complement factor B)
Clonality: Monoclonal
Clone: 10K167
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR