Anti-THY1 Mouse Monoclonal Antibody [clone: 5E10]

Toimittaja: US Biological
C2441-06Q
USBIC2441-06QEA 523 EUR
USBIC2441-06Q
Anti-THY1 Mouse Monoclonal Antibody [clone: 5E10]
Vasta-aineet
Recommended dilution: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: Monoclonal
Clone: 50000000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR