Anti-THY1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: F15-42-1]

Toimittaja: US Biological
C2441-02F2
USBIC2441-02F2EA 822 EUR
USBIC2441-02F2
Anti-THY1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: F15-42-1]
Vasta-aineet
Recommended dilution: Flow Cytometry: 1:10-1:15

Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: Monoclonal
Clone: F15-42-1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR