Anti-Calcitonin Gene Related Peptide Rabbit polyclonal antibody unconjugated

Toimittaja: US Biological
C0115-02J
USBIC0115-02JEA 539 EUR
USBIC0115-02J
Anti-Calcitonin Gene Related Peptide Rabbit polyclonal antibody unconjugated
Vasta-aineet
Recommended dilution: Immunochemistry: 1:200

Type: Primary
Antigen: Calcitonin Gene Related Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR