Anti-ANTXR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
A2296-97C-PE
USBIA2296-97C-PEEA 884 EUR
USBIA2296-97C-PE
Anti-ANTXR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Recommended dilution: FLISA: 1:1000 Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: ANTXR1 (anthrax toxin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR