Anti-ADK Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
A0882-63-PE
USBIA0882-63-PEEA 884 EUR
USBIA0882-63-PE
Anti-ADK Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: ADK (adenosine kinase)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR