Anti-DNAM-1 Rat Monoclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
167643
USBI167643EA 618 EUR
USBI167643
Anti-DNAM-1 Rat Monoclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Do not freeze!

Type: Primary
Antigen: DNAM-1
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR