Anti-Complement 1s Sheep Polyclonal Antibody

Toimittaja: US Biological
137874
USBI137874EA 494 EUR
USBI137874
Anti-Complement 1s Sheep Polyclonal Antibody
Vasta-aineet
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Complement 1s
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR