Anti-Solute Carrier Family 44 Member 2 Rabbit Polyclonal Antibody

Toimittaja: US Biological
136442
USBI136442EA 681 EUR
USBI136442
Anti-Solute Carrier Family 44 Member 2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Solute Carrier Family 44 Member 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR