Anti-MYT1 Mouse Monoclonal Antibody [clone: 2A7]

Toimittaja: US Biological
130105
USBI130105EA 655 EUR
USBI130105
Anti-MYT1 Mouse Monoclonal Antibody [clone: 2A7]
Vasta-aineet
Type: Primary
Antigen: MYT1 (myelin transcription factor 1)
Clonality: monoclonal
Clone: 2A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR