Anti-BOLL Rabbit Polyclonal Antibody

Toimittaja: US Biological
123979
USBI123979EA 696 EUR
USBI123979
Anti-BOLL Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: BOLL (boule-like RNA-binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR