Anti-ZP3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
044404-PE
USBI044404-PEEA 884 EUR
USBI044404-PE
Anti-ZP3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ZP3 (zona pellucida glycoprotein 3 (sperm receptor))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR