Anti-SUPT6H Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Toimittaja: US Biological
042488-APC
USBI042488-APCEA 884 EUR
USBI042488-APC
Anti-SUPT6H Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SUPT6H (suppressor of Ty 6 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR