Anti-PARVA Rabbit Polyclonal Antibody (Biotin)

Toimittaja: US Biological
039689-BIOTIN
USBI039689-BIOTINEA 884 EUR
USBI039689-BIOTIN
Anti-PARVA Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PARVA (parvin, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR