Anti-PAR6G Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
039652-PE
USBI039652-PEEA 884 EUR
USBI039652-PE
Anti-PAR6G Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: PAR6G
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR