Anti-MOAP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
038523-PE
USBI038523-PEEA 884 EUR
USBI038523-PE
Anti-MOAP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MOAP1 (modulator of apoptosis 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR