Anti-MEX3B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Toimittaja: US Biological
038392-AP
USBI038392-APEA 884 EUR
USBI038392-AP
Anti-MEX3B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MEX3B (mex-3 RNA binding family member B)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR