Anti-MAF Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
038056-PE
USBI038056-PEEA 884 EUR
USBI038056-PE
Anti-MAF Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MAF (v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR