Anti-K0746 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
037240-PE
USBI037240-PEEA 903 EUR
USBI037240-PE
Anti-K0746 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Do not freeze!

Type: Primary
Antigen: K0746
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR