Anti-GRIA3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
036289-PE
USBI036289-PEEA 884 EUR
USBI036289-PE
Anti-GRIA3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: GRIA3 (glutamate receptor, ionotropic, AMPA 3)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR