Anti-GNA13 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Toimittaja: US Biological
036123-APC
USBI036123-APCEA 884 EUR
USBI036123-APC
Anti-GNA13 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: GNA13 (guanine nucleotide binding protein (G protein), alpha 13)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR