Anti-FASTK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Toimittaja: US Biological
035523-AP
USBI035523-APEA 884 EUR
USBI035523-AP
Anti-FASTK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: FASTK (Fas-activated serine/threonine kinase)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR