Anti-ERV3-1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: US Biological
035223-PE
USBI035223-PEEA 903 EUR
USBI035223-PE
Anti-ERV3-1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: ERV3-1 (endogenous retrovirus group 3, member 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR