Anti-AMY2B Rabbit Polyclonal Antibody

Toimittaja: US Biological
029718
USBI029718EA 449 EUR
USBI029718
Anti-AMY2B Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: AMY2B (amylase, alpha 2B (pancreatic))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR