Retort rings with bosshead
Toimittaja: USBECK Laborgeräte
2950 2951 2952
USBE2950EA 21.7 EUR
USBE2950 USBE2951 USBE2952
Retort rings with bosshead
Statiivirenkaat
Wire (Ø 4 mm), powder-coated.
With nickel plated brass screw