Burner, Teclu
Toimittaja: USBECK Laborgeräte
1200 1201 1210 1211 1300 1320 1210/1 1211/1 1301
USBE1200EA 75 EUR
USBE1200 USBE1201 451-0086 451-0087 451-1366 451-0088 451-1341 451-1343 USBE1301
Burner, Teclu
Lämmittimet Kaasupolttimet
Nickel plated brass base, in powder coated grey.
  • With air regulation valve and throttle screw
  • Without pilot light
Height: 165 mm