Fittings, VacuTight™, Upchurch Scientific®
Toimittaja: UPCHURCH SCIENTIFIC
P-840 P-844 P-844X P-938 P-940 P-940X P-948
UPCHP-840EA 4.9 EUR
UPCHP-840 UPCHP-844 UPCHP-844X UPCHP-938 UPCHP-940 UPCHP-940X UPCHP-948
Fittings, VacuTight™, Upchurch Scientific®
Kiinnitykset ja liittimet
VacuTight™ fittings provides airtight, dependable connections under vacuum and low pressure conditions. These nuts have streamlined profiles for use in systems requiring a large number of connections in a small area, the ferrules are small size ensures sufficient nut/thread engagement, even in shallow ports. Ideal for combichem, high throughput screening, clinical diagnostic and other automated liquid handling applications.
  • The 10-32 fittings are for use with ¹/₁₆" OD tubing
  • The 1/4-28 and M6 fittings are available for use with both ¹/₁₆" and ¹/₈" OD