VHP Microferrules and nuts, Upchurch Scientific®
Toimittaja: UPCHURCH SCIENTIFIC
PK-152 P-416 P-416BLK PK-112
UPCHPK-152EA 23.6 EUR
UPCHPK-152 UPCHP-416 UPCHP-416BLK UPCHPK-112
VHP Microferrules and nuts, Upchurch Scientific®
Kiinnitykset ja liittimet
VHP Microferrules are made from a proprietary high performance PEEK polymer blend, a material which is unique in its ability to enable the use of capillary tubing in UHPLC environments.
  • The high pressure Microferrules are incorporated into several of VHP products for capillary tubing
  • MicroTight female nuts may be used with any of the separate Microferrules, these ferrules are port-specific and are thusnot interchangeable