Tubing sleeves, FEP, NanoTight™, Upchurch Scientific®
Toimittaja: UPCHURCH SCIENTIFIC
F-237 F-238 F-239 F-239X F-240 F-241 F-242 F-242X F-243 F-244 F-245 F-246 F-247 F-252 F-252X
UPCHF-237EA 7.1 EUR
UPCHF-237 UPCHF-238 UPCHF-239 UPCHF-239X UPCHF-240 UPCHF-241 UPCHF-242 UPCHF-242X UPCHF-243 UPCHF-244 UPCHF-245 UPCHF-246 UPCHF-247 UPCHF-252 UPCHF-252X
Tubing sleeves, FEP, NanoTight™, Upchurch Scientific®
Letku Chromatography Tubing
FEP.
NanoTight tubing sleeves are manufactured using versatile FEP and are precisely cut to a 1,6" length. We offer a wide assortment of sleeves ensuring availability for most applications. Many of the sleeves feature a light colour tint that can help more easily identify the inner diameter for future orders. Because FEP is the base polymer for these sleeves, there is a maximum recommended continuous operating temperature of 50 °C.