Tubing sleeves, PEEK, Upchurch Scientific®
Toimittaja: UPCHURCH SCIENTIFIC
F-227 F-228 F-229 F-230 F-231 F-232 F-233 F-234 F-226 F-225
UPCHF-227EA 9.8 EUR
UPCHF-227 UPCHF-228 UPCHF-229 UPCHF-230 UPCHF-231 UPCHF-232 UPCHF-233 UPCHF-234 UPCHF-226 UPCHF-225
Tubing sleeves, PEEK, Upchurch Scientific®
Letku Chromatography Tubing
PEEK.
The 1/32" O.D. PEEK tubing sleeves is designed to be used with any fitting made for 1/32" O.D. tubing when smaller tubing must be connected. It is 1,6" in length. Select the appropriate sleeve from the product listing for your capillary tubing O.D. size. The 1/32" O.D. PEEK tubing sleeves have a maximum recommended temperature of 125 ºC and have a pressure rating of 5000 psi (345 bar).