Sample loops for high pressure valves, Upchurch Scientific®
Toimittaja: UPCHURCH SCIENTIFIC
1800 1805-1 1810 1826
UPCH1800EA 67.4 EUR
UPCH1800 UPCH1805-1 UPCH1810 UPCH1826
Sample loops for high pressure valves, Upchurch Scientific®
Kromatografiaan tarvikkeet Kromatografiaan näytesilmukat
These PEEK sample loops are metal-free alternatives to stainless steel loops. PEEK loop ends are provided with clean, straight cuts for easy valve installation.
  • PEEK polymer is inert to almost all organic solvents and is biocompatible
  • Flexible, supplied with unswaged fittings but do not require the same swaging precaution
The fittings can reposition along the loop tubing when the fitting reinserts in the ports for correct loop installation.