Autoclaves, T-Edge
Toimittaja: TUTTNAUER
AMS10230WPEDT AMS11230WPEDT TEDGE11230PED
TUTTAMS10230WPEDTEA 7660 EUR
TUTTAMS10230WPEDT TUTTAMS11230WPEDT TUTTTEDGE11230PED
Autoclaves, T-Edge
Sterilisaattorit Autoklaavit (höyrysterilointilaitteet)
The T-Edge 10 and T-Edge 11 are space-saving tabletop autoclaves that give you high speed Class B performance saving you time so you can assist your patients, while meeting current international standards and regulatory requirements to protect your staff and patients.
  • Eco-friendly - Low water consumption per cycle and lower electricity costs
  • Accessible and ergonomic - Convenient and easy loading with door opening to 110°
  • Assorted water filling and drainage: top, front or automatic
  • Designed to use more than 75% of the chamber capacity, with 5 extra-long