Total suspended solids calibration standards, Supelco®
Toimittaja: Merck
TSS100-500ML TSS1000-100ML TSS1000-500ML TSS100-100ML TSS10-100ML TSS10-500ML
TSS100-500MLEA 102 EUR
TSS100-500ML TSS1000-100ML TSS1000-500ML TSS100-100ML TSS10-100ML TSS10-500ML
Total suspended solids calibration standards, Supelco®
Standardiaineet ja vertailumateriaalit Sameustandardit
Total suspended solids (TSS) is the dry-weight of suspended particles, that are not dissolved, in a sample of water that can be trapped by a filter.
  • Useful for waste water analysis
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.