Quartz Glass Rods
Toimittaja: TECHNICAL QUARTZ SOLUTIONS
75350 75351 75352 75353 75354 75355
TSLA75350EA 40.2 EUR
TSLA75350 TSLA75351 TSLA75352 TSLA75353 TSLA75354 TSLA75355
Quartz Glass Rods
Sekoittajat Sekoitussauvat
Temperature resistant glass rods made from transparent quartz glass.
  • Chemical resistance
  • Made for tasks up to 1200 °C
  • High purity
TQS quartz glass rods stand for quality and versatile applications.