Crucibles, low form, quartz
Toimittaja: TECHNICAL QUARTZ SOLUTIONS
75052 75056 75058 75060 75062 75064 75066 75072
TSLA75052EA 27.2 EUR
TSLA75052 TSLA75056 TSLA75058 TSLA75060 TSLA75062 TSLA75064 TSLA75066 TSLA75072
Crucibles, low form, quartz
Upokkaat
Quartz, transparent, glazed.
  • Lids for low form crucible, ring handle
Tilaustiedot: Crucible and lid have to be ordered separately.