Pre-packed columns, 1 and 5 ml, SkillPak™
Toimittaja: TOSOH Bioscience
0045200 0045201 0045202 0045203 0045204 0045205 0045206 0045207 0045208 0045209 0045210 0045211 0045212 0045213 0045214 0045215 0045216 0045217 0045218 0045219 0045220 0045221 0045222 0045223 0045224 0045225 0045226 0045227 0045228 0045229 0045230 0045231 0045232 0045233 0045236 0045239 0045240 0045241 0045242 0045243 0045244 0045245 0045246 0045247 0045248 0045249 0045250 0045251 0045252 0045253 0045254 0045255 0045256 0045257 0045258 0045259 0045260 0045261 0045262 0045263 0045264 0045265 0045266
TOYO0045200EA 168 EUR
TOYO0045200 TOYO0045201 TOYO0045202 TOYO0045203 TOYO0045204 TOYO0045205 TOYO0045206 TOYO0045207 TOYO0045208 TOYO0045209 TOYO0045210 TOYO0045211 TOYO0045212 TOYO0045213 TOYO0045214 TOYO0045215 TOYO0045216 TOYO0045217 TOYO0045218 TOYO0045219 TOYO0045220 TOYO0045221 TOYO0045222 TOYO0045223 TOYO0045224 TOYO0045225 TOYO0045226 TOYO0045227 TOYO0045228 TOYO0045229 TOYO0045230 TOYO0045231 TOYO0045232 TOYO0045233 TOYO0045236 TOYO0045239 TOYO0045240 TOYO0045241 TOYO0045242 TOYO0045243 TOYO0045244 TOYO0045245 TOYO0045246 TOYO0045247 TOYO0045248 TOYO0045249 TOYO0045250 TOYO0045251 TOYO0045252 TOYO0045253 TOYO0045254 TOYO0045255 TOYO0045256 TOYO0045257 TOYO0045258 TOYO0045259 TOYO0045260 TOYO0045261 TOYO0045262 TOYO0045263 TOYO0045264 TOYO0045265 TOYO0045266
Pre-packed columns, 1 and 5 ml, SkillPak™
Kromatografiakolonnit
SkillPak 1 and 5 ml columns are designed for fast method development, resin screening or sample concentration. Pre-packed with TOYOPEARL®, TSKgel® or Ca++Pure-HA™ process chromatography media, these columns offer superior separation of biomelocules, such as monoclonal antibodies, proteins and oligonucleotides. SkillPak 50 and 200 ml pre-packed columns are the best solution for seamless scale up of methods developed on SkillPak 1 and 5.
  • Standard fittings (10-32 for 1/16" capillary)
  • Max. pressure: 3 bar (1 ml columns and 5 ml TOYOPEARL), ≤4 bar (5 ml TSKgel