Ion exchange chromatography sorbents, Toyopearl®, TSK-GEL®
Toimittaja: TOSOH Bioscience
0043280 0043220 0043380
TOYO0043280EA 1250 EUR
TOYO0043280 TOYO0043220 TOYO0043380
Ion exchange chromatography sorbents, Toyopearl®, TSK-GEL®
HPLC- ja TLC, Täytemateriaalit Kromatografiakolonnien täytemateriaalit
Typically they contain three or four different ligand types offered for ion exchange chromatography mode.
The larger resin amounts allow the packing of wider bore and longer columns. This helps more accurate to determine the resin‘s dynamic binding capacity, selectivity, column, efficiency and operating conditions.