Affinity chromatography resins, Toyopearl®, TSK-GEL®
Toimittaja: TOSOH Bioscience
0008000 0008002 0008004 0008006 0008038 0008039 0008040 0008041 0008651 0014471 0014472 0014475 0043411 0019800 0019801 0020030 0016208 0043400 0043402 0043410 0043412 0018827
TOYO0008000EA 418 EUR
TOYO0008000 TOYO0008002 TOYO0008004 TOYO0008006 TOYO0008038 TOYO0008039 TOYO0008040 TOYO0008041 TOYO0008651 TOYO0014471 TOYO0014472 TOYO0014475 TOYO0043411 TOYO0019800 TOYO0019801 TOYO0020030 TOYO0016208 TOYO0043400 TOYO0043402 TOYO0043410 TOYO0043412 TOYO0018827
Affinity chromatography resins, Toyopearl®, TSK-GEL®
HPLC- ja TLC, Täytemateriaalit Kromatografiakolonnien täytemateriaalit
Affinity chromatography resins are functionalised with chemically active groups or group-specific ligands.
  • Standard 100 nm pore size
  • Porous, hydrophilic polymer matrix
  • High mechanical stability
  • High capacity for large biopolymers
It has broad range of applications, suitable for laboratory and process scale purifications, constant bed volume over a wide range of buffer composition.
TOYOPEARL affinity resins may be used in combinatorial chemistry or for solid phase synthesis of peptides and oligonucleotides because of their excellent stability in a variety of organic solvents and under extremes of pH.
Activated resin is highly reactive toward amine and thiol groups. It is provided in dry form, ready for reaction in buffered solutions containing protein or another ligand.
Reactive resins require carbodiimide coupling or reductive amination to achieve a stable covalent linkage.
TSKgel resins are larger particle size versions of the chemically equivalent methacrylic packing of analytical-scale. TSKgel columns used for protein analysis and purification.