Bottles, aluminium, with thread, SYSTEM PLUS® 45
Toimittaja: TOURNAIRE-EMBALLAGE
4829S2 5220S1 7418S1 7419S1
215-3153EA 37.9 EUR
215-3153 TOUR5220S1 TOUR7418S1 TOUR7419S1
Bottles, aluminium, with thread, SYSTEM PLUS® 45
Pullot Metallipullot
Made of pure aluminium.
  • Unbreakable, odorless
  • Very good temperature resistance
  • Leakproof with tamper-evident screw caps (must be odered separately)
  • UN-approved
Suitable for perfumes compositions, aroma chemicals, food ingredients and laboratory liquid reagents.