Vacuum Cleaner, AS-400 MR HEPA (W + D)
Toimittaja: TIGER-VAC
110776B
TIGV110776BEA 1880 EUR
TIGV110776B
Vacuum Cleaner, AS-400 MR HEPA (W + D)
Vakuumipuhdistimet
HEPA filtered vacuum for wet and dry recovery, designed for the recovery of asbestos, lead, mold, etc.
  • Lightweight and portable
  • Heavy duty two-stage by-pass motor
  • Equipped with a three-stage filtration system
  • Middle ring for wet and dry recovery - included
  • Heavy duty plastic recovery tank
  • Stainless steel recovery tank option available
  • Four-wheel cart for easy maneuverability
  • Built-in storage tray with two large 170 mm rear wheels and two 60 mm front wheels
HEPA filter with an efficiency of 99,995% at 0,3 μm. All of our HEPA vacuum systems are aerosolMore Product Information