Vacuum cleaner, B-8 RE HEPA, Tiger-Vac®
Toimittaja: TIGER-VAC
110560B
TIGV110560BEA 1690 EUR
TIGV110560B
Vacuum cleaner, B-8 RE HEPA, Tiger-Vac®
Vakuumipuhdistimet
This unit is designed for general cleanup: for asbestos, silica, lead, and other toxic/nuisance dusts recovery.
  • By-Pass heavy duty motor
  • Equipped with 4 wheels
  • Liquid recovery capacity of 30 liters
  • Silent operation
This unit contains a true HEPA Filter (H14) located downstream outside of the motor. It is equipped with an disposable filter bags for easy and safe disposal of recovered material.
Construction: Polyethylene recovery tank
Varoitus: Note: For wet and dry recovery