Cryogenic Supply Tank, LN₂
Toimittaja: Thermo Fisher Scientific
8120CE 8121CE 8121
THBR8120CEEA 8440 EUR
THBR8120CE THBR8121CE THBR8121
Cryogenic Supply Tank, LN₂
Nestetyppisäiliöt
Transport cryogenic industrial gases with Thermo Scientific™ LN₂ supply tanks, which are designed to be both durable and user-friendly.
  • Rated at 22 psi (151,7 kPa)
  • Evaporate 2% of volume per day
For long-term storage of samples in cryogenic freezers, or controlling temperature during freezing procedures utilizing controlled-rate freezing equipment.